Assay Details
| Gene Symbol: LIX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078003.1 | NP_001071471.1 | 3 - 4 | 526 | 59 | |
| GenBank mRNA | BC126599.1 | - | 3 - 4 | 526 | 59 | |
More Information
Set Membership: |
|

LIX1
Genomic Map