Assay Details
| Gene Symbol: ODAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080315.1 | NP_001073784.1 | 2 - 3 | 115 | 72 | |
| GenBank mRNA | EF121760.1 | - | 2 - 3 | 115 | 72 | |
More Information
Set Membership: |
|

ODAM
Genomic Map