Assay Details
| Gene Symbol: DNAJC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080332.2 | NP_001073801.1 | 1 - 2 | 129 | 90 | |
| GenBank mRNA | BC126836.1 | - | 1 - 2 | 130 | 90 | |
More Information
Set Membership: |
|

DNAJC15
Genomic Map