Assay Details
| Gene Symbol: PI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080353.1 | NP_001073822.1 | 1 - 2 | 100 | 89 | |
| GenBank mRNA | BC126810.1 | - | 1 - 2 | 100 | 89 | |
More Information
Set Membership: |
|

PI3
Genomic Map