Assay Details
| Gene Symbol: AP1AR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081532.1 | NP_001075001.1 | 6 - 7 | 514 | 57 | |
| GenBank mRNA | BC133408.1 | - | 6 - 7 | 514 | 57 | |
| BT030595.1 | - | 6 - 7 | 589 | 57 | ||
More Information
Set Membership: |
|

AP1AR
Genomic Map