Assay Details
| Gene Symbol: CORO1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081590.1 | NP_001075059.1 | 4 - 5 | 530 | 56 | |
| GenBank mRNA | BC133331.1 | - | 4 - 5 | 530 | 56 | |
More Information
Set Membership: |
|

CORO1C
Genomic Map