Assay Details
| Gene Symbol: VIPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081607.1 | NP_001075076.1 | 2 - 3 | 297 | 56 | |
| GenBank mRNA | BC133381.1 | - | 2 - 3 | 297 | 56 | |
More Information
Set Membership: |
|

VIPR1
Genomic Map