Assay Details
| Gene Symbol: TMEM158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081626.1 | NP_001075095.1 | 1 | 1249 | 102 | |
| GenBank mRNA | BC133460.1 | - | 1 | 1249 | 102 | |
More Information
Set Membership: |
|

TMEM158
Genomic Map