Assay Details
| Gene Symbol: KCNAB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081627.1 | NP_001075096.1 | 12 - 13 | 1259 | 65 | |
| GenBank mRNA | BC133385.1 | - | 12 - 13 | 1259 | 65 | |
More Information
Set Membership: |
|

KCNAB3
Genomic Map