Assay Details
| Gene Symbol: GABRD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081715.2 | NP_001075184.1 | 8 - 9 | 1210 | 69 | |
| GenBank mRNA | BC133557.1 | - | 8 - 9 | 1211 | 69 | |
More Information
Set Membership: |
|

GABRD
Genomic Map