Assay Details
| Gene Symbol: ASPHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081741.1 | NP_001075210.1 | 1 - 2 | 1086 | 62 | |
| GenBank mRNA | BC149115.1 | - | 1 - 2 | 926 | 62 | |
| BT029802.1 | - | 1 - 2 | 1086 | 62 | ||
More Information
Set Membership: |
|

ASPHD1
Genomic Map