Assay Details
| Gene Symbol: OCLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082433.2 | NP_001075902.1 | 4 - 5 | 1119 | 83 | |
| GenBank mRNA | AY589500.1 | - | 2 - 3 | 666 | 83 | |
| BC133617.1 | - | 4 - 5 | 1120 | 83 | ||
More Information
Set Membership: |
|

OCLN
Genomic Map