Assay Details
| Gene Symbol: FBXO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082435.2 | NP_001075904.1 | 1 - 2 | 188 | 86 | |
| GenBank mRNA | BC133649.1 | - | 1 - 2 | 189 | 86 | |
More Information
Set Membership: |
|

FBXO5
Genomic Map