Assay Details
| Gene Symbol: CCDC126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082472.1 | NP_001075941.1 | 2 - 3 | 542 | 83 | |
| GenBank mRNA | BC133498.1 | - | 2 - 3 | 542 | 83 | |
More Information
Set Membership: |
|

CCDC126
Genomic Map