Assay Details
| Gene Symbol: SLC22A18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083374.1 | NP_001076843.1 | 3 - 4 | 430 | 60 | |
| GenBank mRNA | BC114890.1 | - | 3 - 4 | 430 | 60 | |
More Information
Set Membership: |
|

SLC22A18
Genomic Map