Assay Details
| Gene Symbol: LYPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083419.1 | NP_001076888.1 | 4 - 5 | 567 | 62 | |
| GenBank mRNA | BC123509.1 | - | 4 - 5 | 567 | 62 | |
More Information
Set Membership: |
|

LYPD3
Genomic Map