Assay Details
| Gene Symbol: STK17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083422.1 | NP_001076891.1 | 6 - 7 | 953 | 96 | |
| GenBank mRNA | BC114806.1 | - | 6 - 7 | 953 | 96 | |
More Information
Set Membership: |
|

STK17A
Genomic Map