Assay Details
| Gene Symbol: LETMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083430.1 | NP_001076899.1 | 4 - 5 | 487 | 71 | |
| GenBank mRNA | BC133596.1 | - | 4 - 5 | 487 | 71 | |
More Information
Set Membership: |
|

LETMD1
Genomic Map