Assay Details
| Gene Symbol: BIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083435.2 | NP_001076904.1 | 8 - 9 | 711 | 89 | |
| GenBank mRNA | BC114869.1 | - | 8 - 9 | 713 | 89 | |
More Information
Set Membership: |
|

BIN2
Genomic Map