Assay Details
| Gene Symbol: GUSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083436.1 | NP_001076905.1 | 11 - 12 | 1833 | 80 | |
| GenBank mRNA | BC133415.1 | - | 11 - 12 | 1833 | 80 | |
More Information
Set Membership: |
|

GUSB
Genomic Map