Assay Details
| Gene Symbol: CIDEA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083449.1 | NP_001076918.1 | 3 - 4 | 372 | 98 | |
| GenBank mRNA | BC123394.1 | - | 3 - 4 | 372 | 98 | |
More Information
Set Membership: |
|

CIDEA
Genomic Map