Assay Details
| Gene Symbol: TMEM38A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083463.1 | NP_001076932.1 | 4 - 5 | 558 | 75 | |
| GenBank mRNA | BC123825.1 | - | 4 - 5 | 558 | 75 | |
More Information
Set Membership: |
|

TMEM38A
Genomic Map