Assay Details
| Gene Symbol: SMYD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083484.1 | NP_001076953.1 | 9 - 10 | 1320 | 59 | |
| GenBank mRNA | BC133278.1 | - | 9 - 10 | 1320 | 59 | |
More Information
Set Membership: |
|

SMYD1
Genomic Map