Assay Details
| Gene Symbol: RHOJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083498.1 | NP_001076967.1 | 3 - 4 | 418 | 76 | |
| GenBank mRNA | BC123637.1 | - | 3 - 4 | 418 | 76 | |
More Information
Set Membership: |
|

RHOJ
Genomic Map