Assay Details
| Gene Symbol: ALG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083511.1 | NP_001076980.1 | 1 - 2 | 203 | 64 | |
| GenBank mRNA | BC123644.1 | - | 1 - 2 | 203 | 64 | |
More Information
Set Membership: |
|

ALG3
Genomic Map