Assay Details
| Gene Symbol: ATG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083531.1 | NP_001077000.1 | 6 - 7 | 593 | 85 | |
| GenBank mRNA | BC114766.1 | - | 6 - 7 | 593 | 85 | |
More Information
Set Membership: |
|

ATG10
Genomic Map