Assay Details
| Gene Symbol: ATP6V1C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083653.1 | NP_001077122.1 | 7 - 8 | 648 | 78 | |
| GenBank mRNA | BC134614.1 | - | 7 - 8 | 648 | 78 | |
More Information
Set Membership: |
|

ATP6V1C2
Genomic Map