Assay Details
| Gene Symbol: ARF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083671.1 | NP_001077140.1 | 4 - 5 | 453 | 61 | |
| GenBank mRNA | BC134695.1 | - | 4 - 5 | 453 | 61 | |
More Information
Set Membership: |
|

ARF5
Genomic Map