Assay Details
| Gene Symbol: TCF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083684.1 | NP_001077153.1 | 2 - 3 | 669 | 83 | |
| GenBank mRNA | BC134562.1 | - | 2 - 3 | 669 | 83 | |
More Information
Set Membership: |
|

TCF19
Genomic Map