Assay Details
| Gene Symbol: TDG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083696.2 | NP_001077165.1 | 4 - 5 | 623 | 59 | |
| GenBank mRNA | BC134451.1 | - | 4 - 5 | 624 | 59 | |
More Information
Set Membership: |
|

TDG
Genomic Map