Assay Details
| Gene Symbol: METTL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083714.1 | NP_001077183.1 | 9 - 10 | 976 | 87 | |
| GenBank mRNA | BC134533.1 | - | 9 - 10 | 976 | 87 | |
More Information
Set Membership: |
|

METTL14
Genomic Map