Assay Details
| Gene Symbol: MYLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083719.1 | NP_001077188.1 | 2 - 3 | 156 | 63 | |
| GenBank mRNA | BC134664.1 | - | 2 - 3 | 156 | 63 | |
More Information
Set Membership: |
|

MYLK2
Genomic Map