Assay Details
| Gene Symbol: SNX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083731.1 | NP_001077200.1 | 1 - 2 | 1757 | 71 | |
| GenBank mRNA | BC134677.1 | - | 1 - 2 | 1757 | 71 | |
More Information
Set Membership: |
|

SNX18
Genomic Map