Assay Details
| Gene Symbol: ZNF689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083746.2 | NP_001077215.1 | 2 - 3 | 661 | 86 | |
| GenBank mRNA | BC134723.1 | - | 2 - 3 | 663 | 86 | |
More Information
Set Membership: |
|

ZNF689
Genomic Map