Assay Details
| Gene Symbol: MYL12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083764.2 | NP_001077233.1 | 1 - 2 | 112 | 147 | |
| GenBank mRNA | BC134471.1 | - | 1 - 2 | 113 | 147 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC134471.1, NM_001083764.2 transcripts.
Set Membership: |
|

MYL12B
Genomic Map