Assay Details
| Gene Symbol: SLPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098865.2 | NP_001092335.1 | 2 - 3 | 248 | 61 | |
| GenBank mRNA | BC146112.1 | - | 2 - 3 | 249 | 61 | |
More Information
Set Membership: |
|

SLPI
Genomic Map