Assay Details
| Gene Symbol: VEGFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101043.2 | NP_001094513.1 | 4 - 5 | 1085 | 88 | |
| GenBank mRNA | AF099135.1 | - | 1 - 2 | 147 | 88 | |
| BC151466.1 | - | 4 - 5 | 1087 | 88 | ||
More Information
Set Membership: |
|

VEGFD
Genomic Map