Assay Details
| Gene Symbol: ADNP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101840.1 | NP_001095310.1 | 2 - 3 | 228 | 68 | |
| GenBank mRNA | BC147927.1 | - | 2 - 3 | 228 | 68 | |
More Information
Set Membership: |
|

ADNP2
Genomic Map