Assay Details
| Gene Symbol: TMEM43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102480.1 | NP_001095950.1 | 11 - 12 | 1192 | 65 | |
| GenBank mRNA | BC149964.1 | - | 11 - 12 | 1192 | 65 | |
More Information
Set Membership: |
|

TMEM43
Genomic Map