Assay Details
| Gene Symbol: BMPR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105328.1 | NP_001098798.1 | 7 - 8 | 1085 | 65 | |
| GenBank mRNA | BC134547.1 | - | 7 - 8 | 1085 | 65 | |
| DQ489533.1 | - | 2 - 3 | 473 | 65 | ||
More Information
Set Membership: |
|

BMPR1B
Genomic Map