Assay Details
| Gene Symbol: GPX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174076.3 | NP_776501.1 | 1 - 2 | 346 | 63 | |
| GenBank mRNA | CK969193.1 | - | 1 - 2 | 597 | 63 | |
| EE243568.1 | - | 1 - 2 | 346 | 63 | ||
| X13684.1 | - | 289 | 63 | |||
More Information
Set Membership: |
|

GPX1
Genomic Map