Assay Details
| Gene Symbol: RDH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174166.3 | NP_776591.2 | 3 - 4 | 681 | 59 | |
| GenBank mRNA | EV670727.1 | - | 3 - 4 | 683 | 59 | |
| EV737918.1 | - | 2 - 3 | 529 | 59 | ||
| L36533.1 | - | 3 - 4 | 682 | 59 | ||
| X82262.1 | - | 610 | 59 | |||
More Information
Set Membership: |
|

RDH5
Genomic Map