Assay Details
| Gene Symbol: RAP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174448.5 | NP_776873.1 | 5 - 6 | 470 | 74 | |
| GenBank mRNA | BC134566.1 | - | 5 - 6 | 400 | 74 | |
| CN789028.1 | - | 5 - 6 | 470 | 74 | ||
| J04196.1 | - | 4 - 5 | 323 | 74 | ||
More Information
Set Membership: |
|

RAP1A
Genomic Map