Assay Details
| Gene Symbol: NAXE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205796.3 | NP_991365.1 | 4 - 5 | 549 | 63 | |
| GenBank mRNA | AY528250.1 | - | 4 - 5 | 550 | 63 | |
| BC114830.1 | - | 4 - 5 | 550 | 63 | ||
More Information
Set Membership: |
|

NAXE
Genomic Map