Assay Details
| Gene Symbol: DOK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024543.2 | NP_001019714.2 | 1 - 2 | 84 | 71 | |
| GenBank mRNA | BT020685.1 | - | 1 - 2 | 84 | 71 | |
| BT021556.1 | - | 1 - 2 | 66 | 71 | ||
| BT029827.1 | - | 1 - 2 | 74 | 71 | ||
More Information
Set Membership: |
|

DOK1
Genomic Map