Assay Details
| Gene Symbol: JMJD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034320.2 | NP_001029492.2 | 4 - 5 | 1097 | 81 | |
| GenBank mRNA | BT021521.1 | - | 4 - 5 | 1097 | 81 | |
More Information
Set Membership: |
|

JMJD6
Genomic Map