Assay Details
| Gene Symbol: SQOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040511.2 | NP_001035601.2 | 4 - 5 | 500 | 123 | |
| GenBank mRNA | BC104605.1 | - | 4 - 5 | 500 | 123 | |
| BT021504.1 | - | 4 - 5 | 123 | |||
More Information
Set Membership: |
|

SQOR
Genomic Map