Assay Details
| Gene Symbol: TUBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075470.2 | NP_001068938.2 | 3 - 4 | 442 | 70 | |
| GenBank mRNA | DV800980.1 | - | 3 - 4 | 470 | 70 | |
More Information
Set Membership: |
|

TUBD1
Genomic Map