Assay Details
| Gene Symbol: DNAJC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075701.2 | NP_001069169.2 | 1 - 2 | 61 | 71 | |
| GenBank mRNA | BC118149.1 | - | 1 - 2 | 61 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001075701.2 transcripts.
Set Membership: |
|

DNAJC22
Genomic Map