Assay Details
| Gene Symbol: EPHX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075855.3 | NP_001069323.2 | 1 - 2 | 311 | 88 | |
| NM_001314056.1 | NP_001300985.1 | 1 - 2 | 311 | 88 | ||
| NR_132766.1 | 1 - 2 | 311 | 88 | |||
| GenBank mRNA | BC120438.1 | - | 1 - 2 | 297 | 88 | |
| DT720129.1 | - | 1 - 2 | 312 | 88 | ||
| DT720346.1 | - | 1 - 2 | 146 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001314056.1 transcripts.
Set Membership: |
|

EPHX4
Genomic Map